4-Hydrazinobenzoesyre 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
411790250
411791000
411790050
ACRO411790250EA
759
DKK
ACRO411790250
ACRO411791000
ACRO411790050
4-Hydrazinobenzoesyre 98%
4-Hydrazinobenzoesyre
Formel:
C₇H₈N₂O₂ Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007581 CAS-nummer: 619-67-0 EINECS: 210-609-5 |
Specifikationer Testresultater
Appearance | Light yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Assay | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...