(Triisopropylsilyl)acetylen 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
368740050
368740250
ACRO368740050EA
945
DKK
ACRO368740050
ACRO368740250
(Triisopropylsilyl)acetylen 97%
(Triisopropylsilyl)acetylen
Formel:
C₁₁H₂₂Si Kogepunkt: 50…52 °C (6 mmHg) Flammepunkt: 56 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00075452 CAS-nummer: 89343-06-6 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4515 to 1.4535 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...