Triethylene glycol dimethyl ether 99% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,5,8,11-Tetraoxadodecane

139600010 139602500 139600025
ACRO139600010EA 1160 DKK
ACRO139600010 ACRO139602500 ACRO139600025
Triethylene glycol dimethyl ether 99% stabilised
Triethylene glycol dimethyl ether
Formel: C₈H₁₈O₄
Kogepunkt: 216 °C (760 mmHg)
Smeltepunkt: –45 °C
Massefylde: 0,986 g/cm³
Flammepunkt: 113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008504
CAS-nummer: 112-49-2
EINECS: 203-977-3

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %
Color scale ≤15 APHA
Stabilizer 0.01 % BHT
Acid value ≤0.05 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR