N-Methylformanilid 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
126810010
126811000
126815000
126810100
ACRO126810010EA
1300
DKK
ACRO126810010
ACRO126811000
ACRO126815000
ACRO126810100
N-Methylformanilid 99%
N-Methylformanilid
Formel:
C₈H₉NO Molekylvægt: 135.17 g/mol Kogepunkt: 243…244 °C (760 mmHg) Smeltepunkt: 8…13 °C Massefylde: 1.09 g/cm³ Flammepunkt: 126 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003283 CAS-nummer: 93-61-8 EINECS: 202-262-3 |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5600 to 1.5620 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...