Dimethylacetylendicarboxylat 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
115710050
115710250
115711000
115715000
ACRO115710050EA
216
DKK
ACRO115710050
ACRO115710250
ACRO115711000
ACRO115715000
Dimethylacetylendicarboxylat 98%
Dimethylacetylendicarboxylat
Formel:
C₆H₆O₄ Kogepunkt: 95…98 °C (19 mmHg) Smeltepunkt: –18 °C Massefylde: 1,16 g/cm³ Flammepunkt: 86 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008456 CAS-nummer: 762-42-5 EINECS: 212-098-4 UN nummer: 3265 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4460 to 1.4500 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...