Calciumperoxid 65%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

21157.22 21157.36
21157.22EA 882 DKK
21157.22 21157.36
Calciumperoxid 65%
Calcium peroxide

CAS No.: 1305-79-9 Molecular Formula: CaO2 Formula Weight: 72.08 Physical Form: Powder Sensitivity: Moisture Sensitive Odour: Odourless Merck Reference: 14,1689 UN No.: 1457 MDL No.: MFCD00015988


Rubber stabilizer

Formel: CaO₂
Molekylvægt: 72.08 g/mol
Smeltepunkt: 692
Massefylde: 2.919…2.921 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015988
CAS-nummer: 1305-79-9
EINECS: 215-139-4
UN nummer: 1457
ADR: 5.1 II
Merck Index: 12,01736

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR