Bariumperoxid 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
350410250
350415000
350410025
ACRO350410250EA
229
DKK
ACRO350410250
ACRO350415000
ACRO350410025
Bariumperoxid 95%
Barium peroxide
Formel:
BaO₂ Molekylvægt: 169.33 g/mol Smeltepunkt: 450 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003454 CAS-nummer: 1304-29-6 EINECS: 215-128-4 UN nummer: 1449 ADR: 5.1 II Merck Index: 12,01018 |
Specifikationer Testresultater
Appearance | Beige Powder |
Titration Iodometric | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...