Svovlsyre 95,0 - 98,0% 95.0-98.0% ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
33273.A1
33273.D6
33273.A1EA
797
DKK
33273.A1
33273.D6
Svovlsyre 95,0 - 98,0% 95.0-98.0% ACS
Svovlsyre
Purity: 95.0-98.0%
CAS No.: 7664-93-9
Molecular Formula: H2SO4
Formula Weight: 98.08
Physical Form: Liquid
Density: 1.84
Appearance: Colourless to pale yellow
Odour: Acrid
Merck Reference: 14,8974
UN No.: 1830
MDL No.: MFCD00064589
Formel:
H₂SO₄ Kogepunkt: ∼335 °C (1013 hPa) Smeltepunkt: 3 °C Massefylde: 1,84 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064589 CAS-nummer: 7664-93-9 EINECS: 231-639-5 UN nummer: 1830 ADR: 8,II Merck Index: 14,8974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...