Natriumsulfit ≥98.0% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

13454.36 13454.A3 13454.A9
13454.36EA 539 DKK
13454.36 13454.A3 13454.A9
Natriumsulfit ≥98.0% ACS
Sodium sulphite

Purity: 98.0% min CAS No.: 7757-83-7 Molecular Formula: Na2SO3 Formula Weight: 126.04 Physical Form: Granular Density: 2.633 Appearance: White Melting Point: 500° dec. Solubility: Soluble in water, glycerol. Practically insoluble in alcohol Merck Reference: 14,8682 MDL No.: MFCD00003503


Photographic developers, generating SO2, reducing agent (dyes), textile bleaching (antichlor)

Formel: Na₂SO₃
Smeltepunkt: >500 °C (dec.)
Massefylde: 2,633
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003503
CAS-nummer: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 14,8682

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR