Phosphonsyre 98%, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Phosphonic acid

201150020 201155000 201150010 201150100
ACRO201150020EA 1840 DKK
ACRO201150020 ACRO201155000 ACRO201150010 ACRO201150100
Phosphonsyre 98%, extra pure
Phosphonsyre
Formel: H₃O₃P
Molekylvægt: 82 g/mol
Smeltepunkt: 73 °C
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00137258
CAS-nummer: 13598-36-2
EINECS: 237-066-7
UN nummer: 2834
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance White Crystals
Titration with NaOH 97.5 to 102.5 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR