Safety shoes for ladies, slip-on, Temptation® City Easy
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6551140/37 6551140/39
DALL6551140/37EA 0 DKK
DALL6551140/37 DALL6551140/39
Safety shoes for ladies, slip-on, Temptation® City Easy
Sko og støvler
These elegant ladies slip-on shoes are made from full grain leather, leather lining plus a PU outsole.
  • Leather lining and leather insole provide comfort and durability
  • Flexible, light sole, slip resistant on different types of surfaces
  • Protective steel toecap (200 J)
Suitable for women working in services and utilities.
Colour: Dark blue.
Certificeringer: EN ISO 20345:2007; S2, SRC.