Multi-layer cell culture flasks, Falcon® Multi-Flask
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
353144 353143
734-2457EA 2310 DKK
734-2457 734-2456
Multi-layer cell culture flasks, Falcon® Multi-Flask
Kolber Celledyrkningskolber
PS, tissue culture-treated, with membrane filter cap, non-pyrogenic.
  • Even distribution of the media across all layers, proven vacuum-gas tissue culture surface treatment and effective gas exchange all combine to provide an optimal cell culture environment
  • Cells can be easily accessed by pipette
  • Batch number printed on each individual flask for improved traceability
  • Manufactured in compliance with cGMP standards
Cell culture treated Falcon® Multi-Flasks are available with 3 or 5 layers and cell growth surfaces 525 or 875 cm². They provide greater surface area and cell yield than single layer T-flasks but with the same footprint and seeding densities, making it easy to scale-up existing protocols.
Graduation accuracy: 10%.
Advarsel: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.