Distributors for bottles
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F746-04 F745-12 F746-12 F746-10 F746-02 F745-10 F745-04 F745-02
BOHLF746-04EA 943 DKK
BOHLF746-04 BOHLF745-12 BOHLF746-12 BOHLF746-10 BOHLF746-02 BOHLF745-10 BOHLF745-04 BOHLF745-02
Distributors for bottles
Låg Flaskelåg
Black screw cap made of PPS and available with or without stopcocks. Distributor body made of PTFE with 2 or 3 ports with female thread UNF 1/4" 28 G on upper and lower sides and stopcock made of FEP for each port.
  • Very good chemical resistance
  • Temperature resistant from −50 to +100 °C
  • Suitable for HPLC applications
The body of the distributor can be turned independently from the screw cap. The completely assembled distributor can be removed and fixed on another bottle without the risk of disarranging the tubing.