Screw caps for glass bottles, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
239246 239236 242217 242210 239207 239494 239239 239240 239241 239242 239243 239244 239273 239274 239275 239211 239213 239214 239216 239217 239218 239238 239220 239237 239222 239225 239226 239227 239229 239231 239233 239235 239284 239281 239425 239433 239443 239446 239401 239285 239286 239287 239288 239289 239290 239291 239292 239283 242222 242224 242225 242228 239209 239201 239203
WHEA239246EA 4390 DKK
WHEA239246 WHEA239236 WHEA242217 WHEA242210 WHEA239207 WHEA239494 WHEA239239 WHEA239240 WHEA239241 WHEA239242 WHEA239243 WHEA239244 WHEA239273 WHEA239274 WHEA239275 WHEA239211 WHEA239213 WHEA239214 WHEA239216 WHEA239217 WHEA239218 WHEA239238 WHEA239220 WHEA239237 WHEA239222 WHEA239225 WHEA239226 WHEA239227 WHEA239229 WHEA239231 WHEA239233 WHEA239235 WHEA239284 WHEA239281 WHEA239425 WHEA239433 WHEA239443 WHEA239446 WHEA239401 WHEA239285 WHEA239286 WHEA239287 WHEA239288 WHEA239289 WHEA239290 WHEA239291 WHEA239292 WHEA239283 WHEA242222 WHEA242224 WHEA242225 WHEA242228 WHEA239209 WHEA239201 WHEA239203
Screw caps for glass bottles, WHEATON®
Låg Flaskelåg
Solid top screw caps made of PP with liners. White polypropylene screw cap provides clear surface for writing on cap.
  • Poly-vinyl liner: Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Use these caps with WHEATON or other manufacturers# glass bottles or vials