Celletællere, Fuchs-Rosenthal
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0640411 0640430 0640431
MARI0640411EA 754 DKK
MARI0640411 MARI0640430 MARI0640431
Celletællere, Fuchs-Rosenthal
Objektglas Tællekamre
These counting chambers with double net ruling are suitable for blood tests in vitro diagnostic (IVD) applications.
Certificeringer: IVD directive 98/79/EC
Leveringsinformation: Supplied with two cover glasses.