Celletællere, Neubauer
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0640111 0640110 0640130 0640131
MARI0640111EA 754 DKK
MARI0640111 MARI0640110 MARI0640130 MARI0640131
Celletællere, Neubauer
Objektglas Tællekamre
These counting chambers are designed for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
Certificeringer: IVD 98/79/EC
Leveringsinformation: Supplied with two cover glasses.