Microscopy slides with 45° ground corners
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1000812 1000912
MARI1000812EA 63.7 DKK
MARI1000812 MARI1000912
Microscopy slides with 45° ground corners
Objektglas Objektglas
Microscopy slides with chamfered corners to reduce the risk of injuries and a silky area of 20mm, on one end, at both sides.
Certificeringer: IVD 98/79/EC