Kaliumnitrit ≥96.0% ACS
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 96.0% min
CAS No.: 7758-09-0
Molecular Formula: KNO2
Formula Weight: 85.11
Physical Form: Solid
Density: 1.915
Appearance: White to pale yellow
Odour: Odourless
pH: 7.0-10.0
Melting Point: 350° dec.
Solubility: Soluble in water, hot alcohol, liquid NH3
Merck Reference: 14,7649
UN No.: 1488
MDL No.: MFCD00011408
In analytical chemistry (testing for amino acids, cobalt, iodine, urea)
Formel:
KNO₂ Smeltepunkt: 350 °C (dec.) Massefylde: 1,915 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011408 CAS-nummer: 7758-09-0 EINECS: 231-832-4 UN nummer: 1488 ADR: 5.1,II Merck Index: 14,7649 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...