Periodsyre for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Orthoperiodsyre
M-Clarity™ quality level = MQ200
8.22288.0100
8.22288.0500
8.22288.0100EA
846
DKK
8.22288.0100
8.22288.0500
Periodsyre for synthesis, Sigma-Aldrich®
Periodsyre
Formel:
H₅IO₆ Molekylvægt: 227.94 g/mol Smeltepunkt: 122 Massefylde: 1.3875 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011344 CAS-nummer: 10450-60-9 EINECS: 233-937-0 UN nummer: 3085 ADR: 5.1 (8) I Merck Index: 13,07248 |
|
Specifikationer Testresultater
Solubility | 2430 g/l (20 °C) |
Melting Point | 122 °C |
Molar Mass | 227.94 g/mol |
Bulk Density | 1400 kg/m3 |
pH | 1.2 (100 g/l, H2O, 20 °C) |
Hazard Class | 5.1 |
Hazard Class Description | Oxidizer |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...