Cover glasses
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0107052 0107222 0107032 0107242 01079990935
630-2186EA 2780 DKK
630-2186 630-2187 630-2185 MARI0107242 MARI01079990935
Cover glasses
Dækglas
Cover slides suitable for fluorescence microscopy.
  • Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.