Flasks, round bottom with ground joint neck, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A158-08 A158-09 A158-06
BOHLA158-08EA 1850 DKK
BOHLA158-08 BOHLA158-09 BOHLA158-06
Flasks, round bottom with ground joint neck, BOLA
Kolber Kolber med rund bund
PFA, transparent.
  • Very good chemical resistance
  • Excellent thermal resistance from −200 to +250 °C
Suitable for vacuum.