Erlenmeyer flasks with ground joints, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A151-03 A151-01 A151-04 A151-02
BOHLA151-03EA 4070 DKK
BOHLA151-03 BOHLA151-01 BOHLA151-04 BOHLA151-02
Erlenmeyer flasks with ground joints, BOLA
Kolber Erlenmeyer-kolber
PTFE, white.
  • Temperature resistance: from −200 to +250°C
  • Universal chemical resistance
  • Vacuum: suitable
  • FDA conform
Thick-walled, with ground joint.