Kaliumchlorat ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

31247-25KG 31247-500G
31247-25KGEA 11120 DKK
31247-25KG 31247-500G
Kaliumchlorat ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™
Potassium chlorate
Formel: KClO₃
Molekylvægt: 122.55 g/mol
Kogepunkt: 400 Dec. (on heating)
Smeltepunkt: 356 °C
Massefylde: 2.34 g/cm³ (23 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011361
CAS-nummer: 3811-04-9
EINECS: 223-289-7
UN nummer: 1485
ADR: 5.1 II
Merck Index: 13,07703

Bestil nu

Specifikationer Testresultater

Assay Min. 99 %
Water insoluble matter Max. 0.005 %
Arsenic (As) Max. 0.00005 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 0.0003 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.01 %
Heavy metals (as Pb) Max. 0.0005 %
Bromate (BrO₃) Max. 0.015 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR