Mechanical balances, Harvard Trip®, 1400 and 1500 series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80000004 80000005 80000003
611-5070EA 4720 DKK
611-5070 611-5089 611-5174
Mechanical balances, Harvard Trip®, 1400 and 1500 series
Vægte
These two pan balances are designed to provide the accuracy necessary in comparative weighing.
Metal base and beam and pan made of stainless steel. Features built-in sliding masses, magnetic damping, self-aligning beam design and floating agate bearings, counterbalancing knob for quick zeroing, hardened steel edges.
Dimensions H×L×W: 160×240×140 mm
Bestillingsoplysninger: Weight set 2000 g model and accessory sold separately.