Magnetic stirring bars, PTFE, cross-shaped, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C369-19 C369-38 C369-32 C369-25 C369-10
BOHLC369-19EA 43.1 DKK
BOHLC369-19 BOHLC369-38 BOHLC369-32 BOHLC369-25 BOHLC369-10
Magnetic stirring bars, PTFE, cross-shaped, BOLA
Omrørere Omrøremagneter
Cross shaped magnetic stirrer bars with magnetic core of Alnico 5 magnet encapsulated in PTFE. Due to stable position optimum stirring can be achieved.
  • Chemical resistant
  • Safe and quiet mixing
Temperature resistance: −200 to +250 °C.