Natriumchlorid
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Saline, NaCl, Sea salt, Halite
P029.1
P029.2
P029.3
ROTHP029.1EA
100.77
DKK
ROTHP029.1
ROTHP029.2
ROTHP029.3
Natriumchlorid
Sodium chloride
Formel:
NaCl Molekylvægt: 58.44 g/mol Kogepunkt: 1413 Smeltepunkt: 801 Massefylde: 2.16 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003477 CAS-nummer: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...