Natriumchlorid
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Saline, NaCl, Sea salt, Halite
9265.1
9265.2
9265.3
9265.4
ROTH9265.1EA
0
DKK
ROTH9265.1
ROTH9265.2
ROTH9265.3
ROTH9265.4
Natriumchlorid
Sodium chloride
Formel:
NaCl Kogepunkt: 1413 °C (1013 hPa) Smeltepunkt: 801 °C Massefylde: 2,16 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003477 CAS-nummer: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...