Kaliumhydrogenkarbonat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassium bicarbonate

M-Clarity™ quality level = MQ300

1.04854.0500 1.04854.9025
1.04854.0500EA 987 DKK
1.04854.0500 1.04854.9025
Kaliumhydrogenkarbonat, EMSURE® for analysis, Supelco®
Kaliumhydrogenkarbonat
Formel: KHCO₃
Molekylvægt: 100.12 g/mol
Smeltepunkt: 292
Massefylde: 2.17 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011402
CAS-nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Bestil nu

Specifikationer Testresultater

Solubility 333 g/l (20 °C)
Molar Mass 100.12 g/mol
Bulk Density 900 - 1100 kg/m3
Density 2.17 g/cm3 (20 °C)
pH 8.0 - 8.6 (50 g/l, H2O, 20 °C)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR