Kaliumhydrogenkarbonat 99.7-100.5% (dried basis) ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Potassium bicarbonate
40195.22
40195.36
40195.22EA
276
DKK
40195.22
40195.36
Kaliumhydrogenkarbonat 99.7-100.5% (dried basis) ACS
Kaliumhydrogenkarbonat
Purity: 99.7-100.5%
CAS No.: 298-14-6
Molecular Formula: KHCO3
Formula Weight: 100.12
Physical Form: Granular
Density: 2.17
Solubility: Slightly soluble in water. Practically insoluble in alcohol
Merck Reference: 14,7609
MDL No.: MFCD00011402
Formel:
KHCO₃ Smeltepunkt: 292 °C Massefylde: 2,17 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011402 CAS-nummer: 298-14-6 EINECS: 206-059-0 Merck Index: 14,7609 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...