Natriumcarbonat, anhydrous, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, NF suitable for use as excipient, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Calcined soda, Carbonic acid disodium salt
1.06398.1000 1.06398.5000 1.06398.9029
1.06398.1000EA 1580 DKK
1.06398.1000 1.06398.5000 1.06398.9029
Natriumcarbonat, anhydrous, EMPROVE® ESSENTIAL BP, Ph. Eur., JP, NF suitable for use as excipient, SAFC®
Sodium carbonate
Formel: Na₂CO₃
Molekylvægt: 105.99 g/mol
Kogepunkt: 1600
Smeltepunkt: 854
Massefylde: 2.53 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003494
CAS-nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM