Magnetic Stirring Bars, PTFE, cylindrical, with pivot ring, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C354-05 C354-11 C354-08 C354-23 C354-35 C354-32 C354-29 C354-26 C354-20 C354-17 C354-14 C354-02
BOHLC354-05EA 24.7 DKK
BOHLC354-05 442-9837 442-9836 442-9838 BOHLC354-35 BOHLC354-32 BOHLC354-29 BOHLC354-26 BOHLC354-20 BOHLC354-17 BOHLC354-14 BOHLC354-02
Magnetic Stirring Bars, PTFE, cylindrical, with pivot ring, BOLA
Omrørere Omrøremagneter
PTFE coated stirring bars. Provide excellent centering and smooth stirring.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications