Tubing connectors, straight
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70877 70878 70879
229-0732EA 185 DKK
229-0732 229-0733 229-0734
Tubing connectors, straight
Fittings og stik
PP, white.
  • Good mechanical strength and chemical resistance
  • Excellent thermal resistance, autoclavable
  • Conform to foodstuff regulations 90/128/CEE, 92/39/CEE and FDA 21 CFR
With tapered ends, for connecting tubing with different inner diameters.