Adapters, bends
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05025004 05025003 05025002 05025001
271-2203EA 335 DKK
271-2203 201-0571 201-0570 LENZ05025001
Adapters, bends
Destillations komponenter Destillationsadaptere
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.