Lithiummetaborat, (dried basis) 99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
A16145.14 A16145.22 A16145.36
A16145.14EA 390 DKK
A16145.14 A16145.22 A16145.36
Lithiummetaborat, (dried basis) 99%
Lithium metaborate
Purity: 99% CAS No.: 13453-69-5 Molecular Formula: BLiO2 Formula Weight: 49.75 Odour: Odourless Melting Point: 845° MDL No.: MFCD00011089
Formel: BLiO₂
Molekylvægt: 49.75 g/mol
Kogepunkt: 1719
Smeltepunkt: 845
Massefylde: 1.397
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011089
CAS-nummer: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546