Cyclohexen stabilised for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M-Clarity™ quality level = MQ200
8.02824.0100
8.02824.0500
8.02824.1000
8.02824.2500
8.02824.0100EA
338
DKK
8.02824.0100
8.02824.0500
8.02824.1000
8.02824.2500
Cyclohexen stabilised for synthesis, Sigma-Aldrich®
Cyclohexene
Formel:
C₆H₁₀ Kogepunkt: 83 °C (1013 hPa) Smeltepunkt: –104 °C Massefylde: 0,81 g/cm³ (20 °C) Flammepunkt: –20 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001539 CAS-nummer: 110-83-8 EINECS: 203-807-8 UN nummer: 2256 ADR: 3,II Merck Index: 13,02756 |
Specifikationer Testresultater
Solubility | 0.21 g/l (20 °C) |
Melting Point | -104 °C |
Molar Mass | 82.14 g/mol |
Boiling Point | 83 °C (1013 hPa) |
Vapor Pressure | 90 hPa (20 °C) |
Flash Point | 1.2 - 7.7 %(V) |
Refractive Index | -20 °C DIN 51755 Part 1 |
Density | 0.81 g/cm3 (20 °C) |
pH | 7 - 8 (0.2 g/l, H2O, 20 °C) |
Ignition Point | 310 °C |
Viscosity kinematic | 0.82 mm2/s (calculated) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...