Brommethyl-cyclopropan 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

221810010 221810050 221810250
ACRO221810010EA 740 DKK
ACRO221810010 ACRO221810050 ACRO221810250
Brommethyl-cyclopropan 96%
Brommethyl-cyclopropan
Formel: C₄H₇Br
Molekylvægt: 135 g/mol
Kogepunkt: 101…108 °C (760 mmHg)
Massefylde: 1.38 g/cm³
Flammepunkt: 41 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00001306
CAS-nummer: 7051-34-5
EINECS: 230-331-8
UN nummer: 1993
ADR: 3 III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥95 %
Specific gravity (20°C) 1.385 to 1.415


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR