Brommethyl-cyclohexan ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L11343.14 L11343.06
L11343.14EA 1130 DKK
L11343.14 L11343.06
Brommethyl-cyclohexan ≥98%
Brommethyl-cyclohexan

Purity: 98% CAS No.: 2550-36-9 Molecular Formula: C7H13Br Formula Weight: 177.09 Flash Point: 68°(154°F) Refractive Index: 1.4935 Boiling Point: 99-100°/40mm Beilstein Reference: 635738 MDL No.: MFCD00001509

Formel: C₇H₁₃Br
Molekylvægt: 177.08 g/mol
Kogepunkt: 99…100 °C (40 mmHg)
Massefylde: 1.269
Flammepunkt: 68 °C (154 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001509
CAS-nummer: 2550-36-9
UN nummer: 1993
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR