Brommethyl-cyclohexan 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
111350050
111350250
111351000
ACRO111350050EA
239
DKK
ACRO111350050
ACRO111350250
ACRO111351000
Brommethyl-cyclohexan 96%
Brommethyl-cyclohexan
Formel:
C₇H₁₃Br Molekylvægt: 177.08 g/mol Kogepunkt: 76…77 °C (26 mmHg) Massefylde: 1.28 g/cm³ Flammepunkt: 57 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001509 CAS-nummer: 2550-36-9 UN nummer: 1993 ADR: 3 III |
Specifikationer Testresultater
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4910 to 1.4930 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...