3-Amino-2-chlor-4-methylpyridin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine

A1855-25G A1855-5G
TCIAA1855-25GEA 1110 DKK
TCIAA1855-25G TCIAA1855-5G
3-Amino-2-chlor-4-methylpyridin ≥98.0% (by GC, titration analysis)
3-Amino-2-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
MDL Number: MFCD00673152
CAS-nummer: 133627-45-9
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f2f9582d01e61b09e9d145d238c5a70f5217100&vid=2f2f9582d01e61b09e9d145d238c5a70f5217100&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR