β-tri-Calciumphosphat (β-fase) ≥98% (β-phase basis), sintered powder, Puriss. p.a. analytical reagent, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

49963-100G 49963-10G
49963-100GEA 6060 DKK
49963-100G 49963-10G
β-tri-Calciumphosphat (β-fase) ≥98% (β-phase basis), sintered powder, Puriss. p.a. analytical reagent, Fluka™
β-tri-Calciumphosphat (β-fase)
Formel: Ca₃O₈P₂
Massefylde: 3,27 g/cm³ (20.5 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015984
CAS-nummer: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741

Bestil nu

Specifikationer Testresultater

Phase-composition (XRD) complying
Arsenic (As) Max. 0.5 ppm
Barium (Ba) Max. 20 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Mercury (Hg) Max. 500 ppb
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 20 ppm
Total S (as SO₄) Max. 500 ppm
Heavy metals (as Pb) Max. 50 ppm
Chloride (Cl) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR