Natriumsaccharinat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1,2-Benzothiazol-3(2H)-one 1,1-dioxide sodium salt

9259.1 9259.2
ROTH9259.1EA 0 DKK
ROTH9259.1 ROTH9259.2
Natriumsaccharinat
Natriumsaccharinat
Formel: C₇H₅NNaO₃S
Molekylvægt: 206.18 g/mol
MDL Number: MFCD00013092
CAS-nummer: 128-44-9
EINECS: 204-886-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR