Natriumhydroxid 2 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Caustic soda
4.80512.9950 4.80512.9010 4.80512.9025 4.80512.1002
4.80512.9950EA 0 DKK
4.80512.9950 4.80512.9010 4.80512.9025 4.80512.1002
Natriumhydroxid 2 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 1390
Smeltepunkt: 323
Massefylde: ≤1.11 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 II
Merck Index: 14,08627