Natriumhydroxid 0.25 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
1.00233.9950 1.00233.9025
1.00233.9950EA 0 DKK
1.00233.9950 1.00233.9025
Natriumhydroxid 0.25 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide
Formel: NaOH
Kogepunkt: ∼105 °C (1013 hPa)
Massefylde: 1,02 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8,II