Zinktrifluoromethansulfonat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate

L15969.06 L15969.14
L15969.06EA 561 DKK
L15969.06 L15969.14
Zinktrifluoromethansulfonat ≥98%
Zinktrifluoromethansulfonat

Purity: 98% CAS No.: 54010-75-2 Molecular Formula: C2F6O6S2Zn Formula Weight: 363.51 Melting Point: 300° Beilstein Reference: 4028195 UN No.: 3261 MDL No.: MFCD00013229

Formel: C₂F₆O₆S₂Zn
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013229
CAS-nummer: 54010-75-2
EINECS: 258-922-6
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR