1-Benzhydrylpiperazin ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
L11530.14
L11530.22
L11530.14EA
323
DKK
L11530.14
L11530.22
1-Benzhydrylpiperazin ≥97%
1-Benzhydrylpiperazin
Purity: 97%
CAS No.: 841-77-0
Molecular Formula: C17H20N2
Formula Weight: 252.36
Melting Point: 91-92°
Beilstein Reference: 222773
Formel:
C₁₇H₂₀N₂ Smeltepunkt: 91…92 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00038379 CAS-nummer: 841-77-0 EINECS: 212-667-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...