Kobber(II)trifluormethansulphonat ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Copper(II) trifluoromethanesulphonate, Copper triflate
B20253.06 B20253.14 B20253.22
B20253.06EA 1060 DKK
B20253.06 B20253.14 B20253.22
Kobber(II)trifluormethansulphonat ≥98%
Kobber(II)trifluormethansulphonat
Purity: 98% CAS No.: 34946-82-2 Molecular Formula: Cu(CF3SO3)2 Formula Weight: 361.68 Melting Point: dec. Beilstein Reference: 4028198 UN No.: 3261 MDL No.: MFCD00077492
Formel: C₂CuF₆O₆S₂
Molekylvægt: 361.69 g/mol
Smeltepunkt: Dec.
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077492
CAS-nummer: 34946-82-2
EINECS: 252-300-8
UN nummer: 3261
ADR: 8 III