Triphenylphosphindibromid 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Dibromotriphenylphosphorane
A11552.09
A11552.18
A11552.30
A11552.09EA
530
DKK
A11552.09
A11552.18
A11552.30
Triphenylphosphindibromid 96%
Triphenylphosphindibromid
Purity: 96%
CAS No.: 1034-39-5
Molecular Formula: (C6H5)3PBr2
Formula Weight: 422.10
Melting Point: ca 260° dec.
Beilstein Reference: 917652
UN No.: 3261
MDL No.: MFCD00000054
Formel:
C₁₈H₁₅Br₂P Molekylvægt: 422.1 g/mol Smeltepunkt: ∼ 260 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000054 CAS-nummer: 1034-39-5 EINECS: 213-855-1 UN nummer: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...