Ammoniumpentaborat octahydrat ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

12294.36 12294.A3
12294.36EA 933 DKK
12294.36 12294.A3
Ammoniumpentaborat octahydrat ≥99%
Ammoniumpentaborat octahydrat

Purity: 99.0% CAS No.: 12046-03-6 Molecular Formula: (NH4)2B10O16.8H2O Formula Weight: 544.29 (400.17anhy) Physical Form: Powder Odour: Odourless MDL No.: MFCD00135839

Formel: H₄B₅NO₈·8H₂O
Molekylvægt: 344.21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00135839
CAS-nummer: 12046-03-6
EINECS: 234-521-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR