3-Methylphthalicanhydrid 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Methylisobenzofuran-1,3-dione

414780050 414780250
ACRO414780050EA 945 DKK
ACRO414780050 ACRO414780250
3-Methylphthalicanhydrid 96%
3-Methylphthalicanhydrid
Formel: C₉H₆O₃
Smeltepunkt: 114…118 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00047316
CAS-nummer: 4792-30-7
EINECS: 225-344-0

Bestil nu

Specifikationer Testresultater

Appearance Yellow to beige to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 114 to 118 °C
Titration with TBAH ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR