Ethyl 3-chlorpropionat 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

118260250 118261000
ACRO118260250EA 326 DKK
ACRO118260250 ACRO118261000
Ethyl 3-chlorpropionat 98%
Ethyl 3-chlorpropionat
Formel: C₅H₉ClO₂
Kogepunkt: 162…163 °C (760 mmHg)
Massefylde: 1,003 g/cm³
Flammepunkt: 54 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000989
CAS-nummer: 623-71-2
EINECS: 210-809-2
UN nummer: 3272
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥97.5 %
Phosphate (PO4) ≤10 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm (by ICP-OES)
Insoluble matter ≤0.01 %
Loss on ignition ≤0.5 %
Nitrogen compounds (as N) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR